Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021958.3 | NP_068777.1 | 1, 2 | 1042 | ||
GenBank mRNA | AK056326.1 | - | 1, 2 | 1039 | ||
AK127923.1 | - | 1, 2 | 932 | |||
AK313298.1 | - | 1, 2 | 929 | |||
BC007294.1 | - | 1, 2 | 1027 | 3349581 | ||
BC033808.1 | - | 1, 2 | 800 | 5184776 | ||
M60721.1 | - | 1, 2 | 998 |