Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134778.1 | NP_001128250.1 | 3 | 459 | ||
NM_006195.5 | NP_006186.1 | 3 | 545 | |||
XM_006717130.1 | XP_006717193.1 | 3 | 511 | |||
XM_006717131.1 | XP_006717194.1 | 3 | 511 | |||
XM_006717132.1 | XP_006717195.1 | 3 | 511 | |||
XM_011518755.1 | XP_011517057.1 | 3 | 459 | |||
XM_017014786.1 | XP_016870275.1 | 3 | 459 | |||
GenBank mRNA | AK092148.1 | - | 3 | 746 | ||
AK299267.1 | - | 2 | 255 | |||
AK309713.1 | - | 3 | 459 | |||
AK312717.1 | - | 3 | 474 | |||
BC016977.1 | - | 1 | 165 | |||
BC094883.1 | - | 3 | 496 | 5212090 | ||
X59841.1 | - | 3 | 541 |