Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042368.2 | NP_001035827.1 | 4, 5 | 653 | ||
NM_001271774.1 | NP_001258703.1 | 4, 5 | 565 | |||
NM_001271775.1 | NP_001258704.1 | 4, 5 | 658 | |||
NM_001271776.1 | NP_001258705.1 | 4, 5 | 638 | |||
NM_006266.3 | NP_006257.1 | 4, 5 | 674 | |||
GenBank mRNA | AF027169.1 | - | 4, 5 | 569 | ||
AF295773.1 | - | 4, 5 | 589 | |||
AK056462.1 | - | 3, 4 | 351 | |||
AK074114.1 | - | 4, 5 | 544 | |||
AK127524.1 | - | 4, 5 | 658 | |||
AK301470.1 | - | 4, 5 | 525 | |||
BC059362.1 | - | 4, 5 | 628 | 30340922 |