Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022064.4 | NP_071347.2 | 20 | 1871 | ||
NR_135218.1 | 20 | 1871 | ||||
XM_006713285.1 | XP_006713348.1 | 20 | 2029 | |||
XM_011533995.1 | XP_011532297.1 | 20 | 1806 | |||
XM_017007018.1 | XP_016862507.1 | 20 | 1833 | |||
GenBank mRNA | AF370367.1 | - | 11 | 1025 | ||
AK026968.1 | - | 2 | 125 | |||
AK090632.1 | - | 10 | 913 | |||
AK096353.1 | - | 20 | 1785 | |||
AK308130.1 | - | 20 | 1813 | |||
AK308607.1 | - | 17 | 1549 | |||
AL832034.1 | - | 20 | 1833 | |||
AL832138.1 | - | 20 | 1808 | |||
AL832344.1 | - | 20 | 1810 | |||
AY744152.1 | - | 19 | 1754 | |||
BC041145.1 | - | 5 | 404 | |||
BC057392.1 | - | 7 | 561 | |||
BC088801.1 | - | 2 | 132 | |||
BC130632.1 | - | 20 | 1812 | 40146663 | ||
CR627051.1 | - | 18 | 2379 |