Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPAS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193314.1 | NP_001180243.1 | 4, 5 | 897 | ||
NM_001193315.1 | NP_001180244.1 | 4, 5 | 497 | |||
NM_001193317.1 | NP_001180246.1 | 4, 5 | 674 | |||
NM_022067.3 | NP_071350.2 | 5, 6 | 768 | |||
XM_011537066.1 | XP_011535368.1 | 4, 5 | 488 | |||
XM_017021579.1 | XP_016877068.1 | 4, 5 | 622 | |||
XM_017021580.1 | XP_016877069.1 | 4, 5 | 488 | |||
XM_017021581.1 | XP_016877070.1 | 4, 5 | 622 | |||
GenBank mRNA | AK022769.1 | - | 4, 5 | 445 | ||
AK022925.1 | - | 4, 5 | 423 | |||
AK098254.1 | - | 4, 5 | 858 | |||
AK222706.1 | - | 4, 5 | 443 | |||
AK297388.1 | - | 4, 5 | 599 | |||
BC015054.1 | - | 5, 6 | 689 | 3920459 | ||
CR457294.1 | - | 3, 4 | 351 | |||
DA794284.1 | - | 4, 5 | 497 |