Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM111A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142519.2 | NP_001135991.1 | 5 | 1968 | ||
NM_001142520.2 | NP_001135992.1 | 5 | 1991 | |||
NM_001142521.2 | NP_001135993.1 | 5 | 1804 | |||
NM_001312909.1 | NP_001299838.1 | 6 | 2206 | |||
NM_001312910.1 | NP_001299839.1 | 5 | 2030 | |||
NM_001312911.1 | NP_001299840.1 | 4 | 1795 | |||
NM_022074.3 | NP_071357.2 | 4 | 1702 | |||
NM_198847.2 | NP_942144.1 | 4 | 1663 | |||
GenBank mRNA | AB067482.1 | - | 2 | 2106 | ||
AK025319.1 | - | 1 | 124 | |||
AK026447.1 | - | 1 | 313 | |||
AK092061.1 | - | 4 | 1646 | |||
AK092953.1 | - | 1 | 882 | |||
AK130799.1 | - | 1 | 23 | |||
AK291453.1 | - | 4 | 1668 | |||
BC013137.1 | - | 1 | 898 | |||
BC054515.1 | - | 1 | 207 | |||
BC071759.1 | - | 4 | 1662 | 6451741 | ||
CR749358.1 | - | 3 | 2309 |