Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302643.1 | NP_001289572.1 | 14 | 1673 | ||
NM_022082.3 | NP_071365.3 | 13 | 1611 | |||
XM_011528978.2 | XP_011527280.1 | 13 | 1523 | |||
GenBank mRNA | AK027065.1 | - | 5 | 1204 | ||
AK074107.1 | - | 11 | 3011 | |||
AK095473.1 | - | 13 | 1462 | |||
AK125597.1 | - | 13 | 2134 | |||
BC025312.1 | - | 14 | 1613 | 4563089 | ||
BC027447.1 | - | 9 | 832 | |||
BC038593.1 | - | 13 | 1453 |