Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318253.1 | NP_001305182.1 | 7 | 977 | ||
NM_133171.4 | NP_573403.1 | 7 | 909 | |||
NM_182764.2 | NP_877496.1 | 6 | 805 | |||
XM_005260496.3 | XP_005260553.1 | 7 | 474 | |||
XM_005260498.3 | XP_005260555.1 | 6 | 836 | |||
XM_005260499.3 | XP_005260556.1 | 6 | 807 | |||
XM_005260500.3 | XP_005260557.1 | 5 | 335 | |||
XM_005260501.4 | XP_005260558.1 | 7 | 445 | |||
XM_006723854.3 | XP_006723917.1 | 6 | 408 | |||
XM_017028009.1 | XP_016883498.1 | 7 | 880 | |||
XM_017028010.1 | XP_016883499.1 | 6 | 383 | |||
XM_017028011.1 | XP_016883500.1 | 6 | 836 | |||
GenBank mRNA | AB058737.1 | - | 6 | 420 | ||
AF398886.1 | - | 5 | 348 | |||
AF417861.1 | - | 5 | 280 | |||
AK057032.1 | - | 7 | 433 | |||
AK293861.1 | - | 6 | 473 | |||
AK299318.1 | - | 8 | 484 | |||
BC000143.4 | - | 6 | 485 | 3047992 | ||
BX648819.1 | - | 6 | 386 |