Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199551.1 | NP_001186480.1 | 3 | 228 | ||
NM_001199552.1 | NP_001186481.1 | 3 | 228 | |||
NM_001199623.1 | NP_001186552.1 | 3 | 231 | |||
NM_001321142.1 | NP_001308071.1 | 4 | 341 | |||
NM_022094.3 | NP_071377.2 | 3 | 312 | |||
GenBank mRNA | AF303893.1 | - | 3 | 312 | ||
AL598630.1 | - | 3 | 232 | |||
AY364638.1 | - | 2 | 174 | |||
BC016851.1 | - | 2 | 207 | 3851667 | ||
BQ695178.1 | - | 4 | 327 | |||
CD518729.1 | - | 2 | 207 | |||
CD518776.1 | - | 3 | 313 | |||
CD519587.1 | - | 2 | 207 |