Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022095.3 | NP_071378.1 | 6 | 1037 | ||
XM_005260504.4 | XP_005260561.1 | 6 | 1750 | |||
XM_011528979.2 | XP_011527281.1 | 5 | 1405 | |||
XM_017028012.1 | XP_016883501.1 | 5 | 808 | |||
GenBank mRNA | AF395833.1 | - | 6 | 1014 | ||
AK054694.1 | - | 2 | 1225 | |||
AK092485.1 | - | 4 | 1246 | |||
AK296989.1 | - | 3 | 608 |