Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022098.3 | NP_071381.1 | 1, 2 | 173 | ||
GenBank mRNA | AK301635.1 | - | 1, 2 | 172 | ||
AK313770.1 | - | 1, 2 | 134 | |||
AL365514.1 | - | 1, 2 | 156 | |||
AL834310.1 | - | 1, 2 | 123 | |||
BC001208.1 | - | 1, 2 | 108 | 3502251 | ||
BC001681.2 | - | 1, 2 | 140 | 2823656 | ||
BC004989.1 | - | 1, 2 | 140 | 2823656 | ||
BP346902.1 | - | 1, 2 | 177 | |||
BX648018.1 | - | 1, 2 | 168 | |||
CR456442.1 | - | 1, 2 | 109 | |||
CR457396.1 | - | 1, 2 | 72 |