Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC28A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199633.1 | NP_001186562.1 | 2 | 251 | ||
NM_022127.2 | NP_071410.1 | 3 | 197 | |||
NR_037638.2 | 2 | 251 | ||||
XM_011518905.2 | XP_011517207.1 | 2 | 237 | |||
XM_011518906.2 | XP_011517208.1 | 3 | 333 | |||
XM_011518907.2 | XP_011517209.1 | 2 | 228 | |||
XM_011518909.2 | XP_011517211.1 | 2 | 237 | |||
XM_011518910.2 | XP_011517212.1 | 2 | 237 | |||
GenBank mRNA | AF305210.1 | - | 3 | 197 | ||
AK292849.1 | - | 2 | 250 | |||
AK304406.1 | - | 2 | 231 | |||
AK310861.1 | - | 2 | 220 | |||
AK314039.1 | - | 2 | 231 | |||
BC093821.1 | - | 2 | 134 | 7939666 | ||
BC093823.1 | - | 2 | 134 | 7939668 | ||
DB218604.1 | - | 2 | 251 | |||
DB237216.1 | - | 2 | 246 |