Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARVG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137605.2 | NP_001131077.1 | 3, 4 | 498 | ||
NM_022141.6 | NP_071424.1 | 3, 4 | 558 | |||
XM_005261702.3 | XP_005261759.1 | 3, 4 | 285 | |||
XM_011530302.2 | XP_011528604.1 | 2, 3 | 3050 | |||
XM_017028907.1 | XP_016884396.1 | 3, 4 | 297 | |||
XM_017028908.1 | XP_016884397.1 | 3, 4 | 317 | |||
GenBank mRNA | AF237772.1 | - | 3, 4 | 352 | ||
AK225929.1 | - | 3, 4 | 501 | |||
AK293360.1 | - | 2, 3 | 279 | |||
AK307294.1 | - | 3, 4 | 497 | |||
AK307299.1 | - | 3, 4 | 297 | |||
AK309348.1 | - | 2, 3 | 294 | |||
AK310087.1 | - | 2, 3 | 114 | |||
AK311264.1 | - | 3, 4 | 283 | |||
AL355092.1 | - | 2, 3 | 265 | |||
AL590887.1 | - | 1, 2 | 74 | |||
BC034406.1 | - | 3, 4 | 479 | 5178529 | ||
BI818662.1 | - | 3, 4 | 305 | |||
BP299653.1 | - | 3, 4 | 499 | |||
BP299720.1 | - | 3, 4 | 324 | |||
CR456480.2 | - | 2, 3 | 129 | |||
DA946316.1 | - | 3, 4 | 317 |