Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021994.2 | NP_068834.2 | 3 | 448 | ||
XM_011515768.2 | XP_011514070.1 | 3 | 420 | |||
GenBank mRNA | AF209198.1 | - | 3 | 448 | ||
AF308819.1 | - | 3 | 319 | |||
AK314630.1 | - | 3 | 336 | |||
AL713791.1 | - | 3 | 337 | |||
BC020626.1 | - | 3 | 335 | 4616168 |