Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022041.3 | NP_071324.1 | 3 | 443 | ||
XM_017023734.1 | XP_016879223.1 | 2 | 353 | |||
GenBank mRNA | AF291673.1 | - | 3 | 439 | ||
AK095378.1 | - | 3 | 443 | |||
BC044840.1 | - | 3 | 430 | 2900666 | ||
KU178409.1 | - | 3 | 295 |