Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022045.4 | NP_071328.2 | 4 | 485 | ||
XM_011516962.2 | XP_011515264.1 | 4 | 482 | |||
XM_011516963.2 | XP_011515265.1 | 4 | 482 | |||
GenBank mRNA | AK022122.1 | - | 4 | 469 | ||
AK300762.1 | - | 4 | 461 | |||
BC013136.1 | - | 4 | 457 | 4476886 |