Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022064.4 | NP_071347.2 | 21 | 1940 | ||
NR_135218.1 | 21 | 1940 | ||||
XM_006713285.1 | XP_006713348.1 | 21 | 2098 | |||
XM_011533995.1 | XP_011532297.1 | 21 | 1875 | |||
XM_017007018.1 | XP_016862507.1 | 21 | 1902 | |||
GenBank mRNA | AF370367.1 | - | 12 | 1094 | ||
AK026968.1 | - | 3 | 194 | |||
AK090632.1 | - | 11 | 982 | |||
AK096353.1 | - | 21 | 1854 | |||
AK126883.1 | - | 7 | 489 | |||
AK308130.1 | - | 21 | 1882 | |||
AK308607.1 | - | 18 | 1618 | |||
AL832034.1 | - | 21 | 1902 | |||
AL832138.1 | - | 21 | 1877 | |||
AL832344.1 | - | 21 | 1879 | |||
AY744152.1 | - | 20 | 1823 | |||
BC041145.1 | - | 6 | 473 | |||
BC057392.1 | - | 8 | 630 | |||
BC088801.1 | - | 3 | 201 | |||
BC130632.1 | - | 21 | 1881 | 40146663 | ||
CR627051.1 | - | 19 | 2448 |