Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318253.1 | NP_001305182.1 | 7 | 1097 | ||
NM_133171.4 | NP_573403.1 | 7 | 1029 | |||
NM_182764.2 | NP_877496.1 | 6 | 925 | |||
XM_005260496.3 | XP_005260553.1 | 7 | 594 | |||
XM_005260498.3 | XP_005260555.1 | 6 | 956 | |||
XM_005260499.3 | XP_005260556.1 | 6 | 927 | |||
XM_005260500.3 | XP_005260557.1 | 5 | 455 | |||
XM_005260501.4 | XP_005260558.1 | 7 | 565 | |||
XM_006723854.3 | XP_006723917.1 | 6 | 528 | |||
XM_017028009.1 | XP_016883498.1 | 7 | 1000 | |||
XM_017028010.1 | XP_016883499.1 | 6 | 503 | |||
XM_017028011.1 | XP_016883500.1 | 6 | 956 | |||
GenBank mRNA | AB058737.1 | - | 6 | 540 | ||
AF398886.1 | - | 5 | 468 | |||
AF417861.1 | - | 5 | 400 | |||
AK057032.1 | - | 7 | 553 | |||
AK293861.1 | - | 6 | 593 | |||
AK299318.1 | - | 8 | 604 | |||
AY947527.1 | - | 1 | 35 | |||
BC000143.4 | - | 6 | 605 | 3047992 | ||
BX648819.1 | - | 6 | 506 |