Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIGNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042762.2 | NP_001036227.1 | 4 | 541 | ||
NM_001287492.1 | NP_001274421.1 | 4 | 979 | |||
NM_001287493.1 | NP_001274422.1 | 3 | 373 | |||
NM_001287494.1 | NP_001274423.1 | 3 | 368 | |||
NM_001287495.1 | NP_001274424.1 | 2 | 264 | |||
NM_022116.4 | NP_071399.2 | 4 | 534 | |||
XM_011515470.2 | XP_011513772.1 | 4 | 543 | |||
XM_017012501.1 | XP_016867990.1 | 4 | 1890 | |||
XM_017012502.1 | XP_016867991.1 | 5 | 1062 | |||
GenBank mRNA | AK023142.1 | - | 4 | 494 | ||
AK027537.1 | - | 4 | 941 | |||
AK308136.1 | - | 3 | 330 | |||
BC051867.1 | - | 4 | 475 | 6158883 | ||
DA121143.1 | - | 2 | 221 | |||
DA121992.1 | - | 3 | 325 | |||
DA433617.1 | - | 3 | 335 | |||
DA812999.1 | - | 3 | 453 |