Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022119.3 | NP_071402.1 | 4 | 483 | ||
XM_005255473.3 | XP_005255530.1 | 5 | 582 | |||
GenBank mRNA | AB010779.1 | - | 4 | 420 | ||
AF321182.1 | - | 4 | 434 | |||
AK309624.1 | - | 4 | 625 | |||
AY358396.1 | - | 4 | 469 | |||
BC009726.1 | - | 4 | 455 | 3899480 | ||
BQ893064.1 | - | 491 |