Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLSTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022131.2 | NP_071414.2 | 3, 4 | 627 | ||
XM_017007022.1 | XP_016862511.1 | 3, 4 | 668 | |||
GenBank mRNA | AJ278018.1 | - | 3, 4 | 447 | ||
AK096432.1 | - | 3, 4 | 627 | |||
AK315308.1 | - | 3, 4 | 627 | |||
AY753303.1 | - | 3, 4 | 437 | |||
BC103496.1 | - | 3, 4 | 447 | 40008317 | ||
BC103506.1 | - | 3, 4 | 446 | 40008319 | ||
BC104485.1 | - | 3, 4 | 444 | 40008318 |