Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POPDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308333.1 | NP_001295262.1 | 1 | 556 | ||
NM_022135.3 | NP_071418.2 | 1 | 556 | |||
XM_005247698.3 | XP_005247755.1 | 1 | 556 | |||
GenBank mRNA | AF204173.1 | - | 1 | 539 | ||
AK226022.1 | - | 2 | 621 | |||
AK377174.1 | - | 5 | 445 | |||
AK377178.1 | - | 4 | 372 | |||
AK377179.1 | - | 4 | 372 | |||
AK377180.1 | - | 5 | 445 | |||
AK377182.1 | - | 4 | 372 | |||
AK377184.1 | - | 5 | 445 | |||
BC044929.1 | - | 1 | 556 | 5191230 |