Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022144.2 | NP_071427.2 | 4, 5 | 645 | ||
GenBank mRNA | AB055421.1 | - | 4, 5 | 425 | ||
AF191770.1 | - | 4, 5 | 605 | |||
AF234259.1 | - | 4, 5 | 486 | |||
AF291656.1 | - | 4, 5 | 491 | |||
AY358706.1 | - | 4, 5 | 477 |