Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022045.4 | NP_071328.2 | 12 | 1300 | ||
XM_011516962.2 | XP_011515264.1 | 12 | 1297 | |||
XM_011516963.2 | XP_011515265.1 | 12 | 1297 | |||
GenBank mRNA | BC013136.1 | - | 12 | 1272 | 4476886 |