Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022064.4 | NP_071347.2 | 26 | 2579 | ||
NR_135218.1 | 26 | 2583 | ||||
XM_006713285.1 | XP_006713348.1 | 26 | 2737 | |||
XM_011533995.1 | XP_011532297.1 | 26 | 2514 | |||
XM_017007018.1 | XP_016862507.1 | 26 | 2541 | |||
GenBank mRNA | AF370367.1 | - | 17 | 1733 | ||
AK026968.1 | - | 8 | 833 | |||
AK090632.1 | - | 16 | 1621 | |||
AK126883.1 | - | 11 | 1430 | |||
AK308607.1 | - | 23 | 2257 | |||
AL832034.1 | - | 26 | 2541 | |||
AL832138.1 | - | 25 | 2747 | |||
AL832344.1 | - | 26 | 2518 | |||
AY744152.1 | - | 25 | 2462 | |||
BC041145.1 | - | 11 | 1112 | |||
BC057392.1 | - | 13 | 1269 | |||
BC088801.1 | - | 8 | 840 | |||
BC130632.1 | - | 26 | 2520 | 40146663 | ||
CR627051.1 | - | 23 | 3295 |