Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022066.3 | NP_071349.3 | 18 | 3862 | ||
XM_005257567.1 | XP_005257624.1 | 18 | 3849 | |||
XM_017024934.1 | XP_016880423.1 | 18 | 3774 | |||
XM_017024935.1 | XP_016880424.1 | 18 | 3771 | |||
GenBank mRNA | AB051521.1 | - | 18 | 3862 | ||
AK000926.1 | - | 1 | 97 | |||
AK022940.1 | - | 5 | 1360 | |||
AK024657.1 | - | 3 | 861 | |||
AK025999.1 | - | 10 | 2341 | |||
AK225412.1 | - | 10 | 2341 | |||
AL832432.1 | - | 11 | 2787 | |||
BC004525.2 | - | 3 | 677 | |||
BC022237.1 | - | 5 | 1380 | |||
BC025977.1 | - | 5 | 1261 | |||
BC036820.1 | - | 13 | 2998 | |||
BC051868.2 | - | 10 | 2205 | 6162673 |