Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318253.1 | NP_001305182.1 | 10, 11 | 1461 | ||
NM_133171.4 | NP_573403.1 | 10, 11 | 1393 | |||
NM_182764.2 | NP_877496.1 | 9, 10 | 1289 | |||
XM_005260496.3 | XP_005260553.1 | 10, 11 | 958 | |||
XM_005260498.3 | XP_005260555.1 | 9, 10 | 1320 | |||
XM_005260499.3 | XP_005260556.1 | 9, 10 | 1291 | |||
XM_005260500.3 | XP_005260557.1 | 8, 9 | 819 | |||
XM_005260501.4 | XP_005260558.1 | 9, 10 | 829 | |||
XM_006723854.3 | XP_006723917.1 | 9, 10 | 892 | |||
XM_017028009.1 | XP_016883498.1 | 9, 10 | 1264 | |||
XM_017028010.1 | XP_016883499.1 | 8, 9 | 767 | |||
XM_017028011.1 | XP_016883500.1 | 8, 9 | 1220 | |||
GenBank mRNA | AB058737.1 | - | 9, 10 | 904 | ||
AF398886.1 | - | 8, 9 | 832 | |||
AF417861.1 | - | 8, 9 | 764 | |||
AK057032.1 | - | 10, 11 | 917 | |||
AK293861.1 | - | 8, 9 | 857 | |||
BC000143.4 | - | 9, 10 | 969 | 3047992 | ||
BX648819.1 | - | 9, 10 | 870 |