Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304514.1 | NP_001291443.1 | 4 | 522 | ||
NM_022087.3 | NP_071370.2 | 5 | 855 | |||
XM_006716082.2 | XP_006716145.1 | 5 | 766 | |||
XM_006716083.2 | XP_006716146.1 | 5 | 734 | |||
XM_006716084.2 | XP_006716147.1 | 5 | 698 | |||
XM_017012499.1 | XP_016867988.1 | 5 | 836 | |||
XM_017012500.1 | XP_016867989.1 | 4 | 712 | |||
GenBank mRNA | AK025287.1 | - | 5 | 689 | ||
AK124934.1 | - | 7 | 1043 | |||
AK296097.1 | - | 4 | 588 | |||
AK298911.1 | - | 4 | 445 | |||
Y12434.3 | - | 4 | 606 |