Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000249.3 | NP_000240.1 | 6 | 665 | ||
NM_001167617.1 | NP_001161089.1 | 6 | 460 | |||
NM_001167618.1 | NP_001161090.1 | 6 | 460 | |||
NM_001258271.1 | NP_001245200.1 | 6 | 665 | |||
NM_001258273.1 | NP_001245202.1 | 5 | 574 | |||
NM_001258274.1 | NP_001245203.1 | 7 | 610 | |||
XM_005265161.1 | XP_005265218.1 | 6 | 555 | |||
XM_005265163.1 | XP_005265220.1 | 6 | 465 | |||
XM_005265164.1 | XP_005265221.1 | 6 | 518 | |||
XM_005265166.1 | XP_005265223.1 | 6 | 473 | |||
XM_011533727.1 | XP_011532029.1 | 6 | 473 | |||
XM_017006449.1 | XP_016861938.1 | 6 | 473 | |||
XM_017006450.1 | XP_016861939.1 | 6 | 518 | |||
XM_017006451.1 | XP_016861940.1 | 6 | 518 | |||
GenBank mRNA | AK222810.1 | - | 6 | 472 | ||
AK295359.1 | - | 6 | 439 | |||
AK298583.1 | - | 6 | 433 | |||
AK302807.1 | - | 8 | 394 | |||
AK311326.1 | - | 6 | 675 | |||
AK311365.1 | - | 6 | 414 | |||
AK312609.1 | - | 6 | 481 | |||
AK316264.1 | - | 5 | 391 | |||
AU127758.1 | - | 6 | 539 | |||
AY517558.1 | - | 6 | 465 | |||
BC006850.1 | - | 6 | 517 | 3451538 | ||
DA517219.1 | - | 6 | 465 | |||
U07343.1 | - | 6 | 488 | |||
U07418.1 | - | 6 | 508 |