Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199551.1 | NP_001186480.1 | 5 | 352 | ||
NM_001199552.1 | NP_001186481.1 | 4 | 322 | |||
NM_001199623.1 | NP_001186552.1 | 4 | 325 | |||
NM_001321142.1 | NP_001308071.1 | 5 | 435 | |||
NM_001321143.1 | NP_001308072.1 | 4 | 281 | |||
NM_001321144.1 | NP_001308073.1 | 3 | 168 | |||
NM_022094.3 | NP_071377.2 | 4 | 406 | |||
GenBank mRNA | AF303893.1 | - | 4 | 406 | ||
AK024524.1 | - | 3 | 165 | |||
AK024530.1 | - | 3 | 165 | |||
AL598630.1 | - | 4 | 326 | |||
AY364640.1 | - | 3 | 252 | |||
BC016851.1 | - | 3 | 301 | 3851667 | ||
BQ695178.1 | - | 5 | 421 | |||
CD518729.1 | - | 4 | 331 | |||
CD518776.1 | - | 4 | 407 | |||
CD519587.1 | - | 3 | 301 |