Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321355.1 | NP_001308284.1 | 6 | 2307 | ||
NM_001321356.1 | NP_001308285.1 | 7 | 2808 | |||
NM_022103.3 | NP_071386.3 | 5 | 2206 | |||
XM_011527208.1 | XP_011525510.1 | 6 | 4452 | |||
XM_011527209.1 | XP_011525511.1 | 5 | 3966 | |||
GenBank mRNA | AK024073.1 | - | 1 | 1569 | ||
AK056508.1 | - | 5 | 2148 | |||
AK096513.1 | - | 7 | 2737 | |||
AK126957.1 | - | 6 | 3598 | |||
BC136388.1 | - | 4 | 2042 | 9020375 | ||
BC150593.1 | - | 4 | 2042 | 9056963 | ||
BX647401.1 | - | 5 | 2179 | |||
CR936605.1 | - | 6 | 2272 |