Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PERP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022121.4 | NP_071404.2 | 3 | 1180 | ||
GenBank mRNA | AF317550.1 | - | 3 | 1069 | ||
AJ251830.1 | - | 3 | 1034 | |||
AK074585.1 | - | 3 | 1104 | |||
AK075082.1 | - | 3 | 1076 | |||
AK093516.1 | - | 4 | 1022 | |||
AK097387.1 | - | 3 | 989 | |||
AK097958.1 | - | 3 | 1094 | |||
AK225907.1 | - | 3 | 1073 | |||
DR422553.1 | - | 1 | 494 |