Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022122.2 | NP_071405.2 | 7 | 945 | ||
XM_011542948.2 | XP_011541250.1 | 6 | 864 | |||
XM_011542949.2 | XP_011541251.1 | 6 | 831 | |||
XM_011542950.2 | XP_011541252.1 | 6 | 691 | |||
XM_017018120.1 | XP_016873609.1 | 7 | 990 | |||
GenBank mRNA | AF195192.1 | - | 7 | 928 | ||
AY358752.1 | - | 7 | 945 | |||
BX102293.1 | - | 3 | 193 |