Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271851.1 | NP_001258780.1 | 1 | 373 | ||
NM_022157.3 | NP_071440.1 | 1 | 373 | |||
GenBank mRNA | AF272035.1 | - | 1 | 201 | ||
AF323609.1 | - | 1 | 196 | |||
AK298788.1 | - | 1 | 269 | |||
AK315791.1 | - | 1 | 298 | |||
BC016668.1 | - | 1 | 218 | 4342448 | ||
BG717489.1 | - | 1 | 381 | |||
BI461665.1 | - | 1 | 324 |