Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MME | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000902.3 | NP_000893.2 | 14, 15 | 1544 | ||
NM_007287.2 | NP_009218.2 | 14, 15 | 1520 | |||
NM_007288.2 | NP_009219.2 | 14, 15 | 1566 | |||
NM_007289.2 | NP_009220.2 | 14, 15 | 1611 | |||
XM_006713646.3 | XP_006713709.1 | 14, 15 | 1558 | |||
XM_006713647.3 | XP_006713710.1 | 14, 15 | 4704 | |||
XM_011512855.2 | XP_011511157.1 | 14, 15 | 1635 | |||
XM_011512856.2 | XP_011511158.1 | 14, 15 | 1562 | |||
XM_011512857.2 | XP_011511159.1 | 14, 15 | 1533 | |||
GenBank mRNA | AK291761.1 | - | 14, 15 | 1635 | ||
BC101632.1 | - | 14, 15 | 1502 | 8069138 | ||
BC101658.1 | - | 14, 15 | 1500 | 8069164 | ||
BC143465.1 | - | 14, 15 | 1502 | 9051973 | ||
BX647134.1 | - | 14, 15 | 1515 | |||
J03779.1 | - | 13, 14 | 1435 | |||
X07166.1 | - | 13, 14 | 1420 | |||
Y00811.1 | - | 14, 15 | 1559 |