Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022168.3 | NP_071451.2 | 1 | 532 | ||
GenBank mRNA | AF095844.1 | - | 1 | 299 | ||
AK292941.1 | - | 2 | 446 | |||
AK314611.1 | - | 1 | 527 | |||
AY017378.1 | - | 1 | 285 | |||
BC046208.1 | - | 1 | 508 | |||
BC078180.1 | - | 1 | 354 | |||
BC111750.1 | - | 1 | 199 | 40008600 | ||
BU902097.1 | - | 1 | 353 |