Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022337.2 | NP_071732.1 | 1, 2 | 293 | ||
XM_017017455.1 | XP_016872944.1 | 1, 2 | 331 | |||
XM_017017456.1 | XP_016872945.1 | 1, 2 | 331 | |||
GenBank mRNA | AF136172.1 | - | 1, 2 | 214 | ||
AF235022.1 | - | 1, 2 | 257 | |||
AL546159.3 | - | 1, 2 | 293 | |||
BC015808.1 | - | 1, 2 | 259 | 3916084 | ||
BT009842.1 | - | 1, 2 | 210 |