Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022346.4 | NP_071741.2 | 2 | 411 | ||
NR_073124.1 | 2 | 411 | ||||
XM_011513876.2 | XP_011512178.1 | 2 | 295 | |||
XM_017008543.1 | XP_016864032.1 | 2 | 295 | |||
XM_017008544.1 | XP_016864033.1 | 2 | 245 | |||
GenBank mRNA | AB013299.1 | - | 2 | 285 | ||
AF235023.1 | - | 2 | 214 | |||
AF331796.1 | - | 2 | 322 | |||
AK022512.1 | - | 2 | 242 | |||
BC068467.1 | - | 2 | 228 | 4821684 | ||
BC101476.1 | - | 2 | 199 | 8068982 | ||
BX474610.1 | - | 2 | 238 | |||
DB451361.1 | - | 2 | 411 |