Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001336.3 | NP_001327.2 | 5 | 836 | ||
GenBank mRNA | AF009923.1 | - | 5 | 680 | ||
AF073890.1 | - | 5 | 806 | |||
AF136273.1 | - | 5 | 835 | |||
AK314931.1 | - | 5 | 842 | |||
BC042168.1 | - | 5 | 819 | 5018854 | ||
BT019915.1 | - | 5 | 710 |