Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022365.3 | NP_071760.2 | 1 | 478 | ||
XM_011519614.2 | XP_011517916.1 | 1 | 525 | |||
XM_017016536.1 | XP_016872025.1 | 1 | 525 | |||
GenBank mRNA | AK027263.1 | - | 1 | 298 | ||
AK308369.1 | - | 1 | 484 | |||
AY225122.1 | - | 1 | 191 | |||
BC110894.1 | - | 1 | 478 | 5164559 |