Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022371.3 | NP_071766.2 | 3 | 744 | ||
GenBank mRNA | AJ299403.1 | - | 3 | 633 | ||
AJ299441.1 | - | 3 | 633 | |||
AK025998.1 | - | 3 | 623 | |||
AK130132.1 | - | 1 | 63 | |||
BC001085.1 | - | 3 | 642 | 2959819 | ||
BC007571.1 | - | 3 | 633 | 3029304 | ||
BC011746.1 | - | 3 | 629 | 3343691 | ||
BC018292.1 | - | 3 | 634 | 3457259 | ||
CR457354.1 | - | 3 | 592 |