Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLST8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199173.1 | NP_001186102.1 | 5 | 747 | ||
NM_001199174.1 | NP_001186103.1 | 5 | 586 | |||
NM_001199175.1 | NP_001186104.1 | 5 | 774 | |||
NM_022372.4 | NP_071767.3 | 5 | 777 | |||
XM_005255475.3 | XP_005255532.2 | 5 | 553 | |||
XM_005255478.2 | XP_005255535.1 | 5 | 758 | |||
XM_005255479.2 | XP_005255536.1 | 5 | 685 | |||
XM_017023548.1 | XP_016879037.1 | 5 | 742 | |||
XM_017023549.1 | XP_016879038.1 | 5 | 483 | |||
XM_017023551.1 | XP_016879040.1 | 5 | 792 | |||
GenBank mRNA | AF195883.1 | - | 2 | 98 | ||
AK021536.1 | - | 5 | 746 | |||
AK022227.1 | - | 5 | 556 | |||
AK098762.1 | - | 5 | 682 | |||
AK302201.1 | - | 4 | 586 | |||
AU121635.1 | - | 5 | 556 | |||
AY223837.1 | - | 4 | 587 | |||
BC001313.2 | - | 5 | 467 | 3454025 | ||
BC017119.1 | - | 4 | 478 | 3910883 | ||
BC052292.1 | - | 4 | 1001 | 5926090 | ||
BC088354.1 | - | 5 | 495 | 6168345 | ||
BI761765.1 | - | 5 | 505 |