Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105079.2 | NP_001098549.2 | 18 | 2462 | ||
XM_011545916.2 | XP_011544218.1 | 17 | 3809 | |||
XM_011545917.2 | XP_011544219.1 | 17 | 3809 | |||
XM_011545919.2 | XP_011544221.1 | 16 | 3772 | |||
GenBank mRNA | AK021680.1 | - | 6 | 653 | ||
AK022551.1 | - | 9 | 867 | |||
AK093045.1 | - | 2 | 1138 | |||
AK096322.1 | - | 10 | 1245 | |||
AK298229.1 | - | 11 | 1025 |