Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022473.2 | NP_071918.1 | 1 | 523 | ||
XM_005254591.2 | XP_005254648.1 | 4 | 430 | |||
XM_005254592.2 | XP_005254649.1 | 4 | 430 | |||
GenBank mRNA | AF205632.1 | - | 1 | 496 | ||
BC029366.1 | - | 1 | 487 | |||
BC058023.1 | - | 1 | 492 | |||
BC070244.1 | - | 1 | 487 | |||
BC111396.1 | - | 1 | 492 | |||
BX649141.1 | - | 4 | 407 |