Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022478.3 | NP_071923.2 | 13 | 2927 | ||
NM_144985.3 | NP_659422.2 | 12 | 2813 | |||
GenBank mRNA | AK025342.1 | - | 5 | 1231 | ||
AL137477.1 | - | 4 | 1386 | |||
AY260900.1 | - | 11 | 2646 | |||
AY260901.1 | - | 12 | 2760 | |||
AY358199.1 | - | 12 | 2769 | |||
BC094810.1 | - | 1 | 691 | |||
BC110867.1 | - | 5 | 1383 |