Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022480.3 | NP_071925.2 | 2 | 1279 | ||
GenBank mRNA | AK022649.1 | - | 1 | 700 | ||
AK056663.1 | - | 3 | 1121 | |||
AK092231.1 | - | 2 | 1241 | |||
AK315541.1 | - | 2 | 1234 | |||
AL136796.1 | - | 2 | 1191 | |||
AY764034.1 | - | 2 | 1261 | |||
BC028100.1 | - | 2 | 1198 | 5242783 |