Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022481.5 | NP_071926.4 | 5, 6 | 986 | ||
XM_005268497.1 | XP_005268554.1 | 5, 6 | 950 | |||
XM_005268498.1 | XP_005268555.1 | 5, 6 | 950 | |||
XM_005268499.1 | XP_005268556.1 | 5, 6 | 924 | |||
XM_005268500.3 | XP_005268557.1 | 5, 6 | 950 | |||
XM_006714792.1 | XP_006714855.1 | 5, 6 | 950 | |||
XM_011537677.2 | XP_011535979.1 | 3, 4 | 405 | |||
GenBank mRNA | AJ310567.1 | - | 4, 5 | 936 | ||
AK091385.1 | - | 5, 6 | 924 | |||
AK290052.1 | - | 5, 6 | 979 | |||
BC025757.1 | - | 4, 5 | 1264 | |||
BC027880.1 | - | 4, 5 | 1253 |