Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303134.1 | NP_001290063.1 | 2 | 637 | ||
NM_022494.2 | NP_071939.1 | 2 | 637 | |||
XM_005270051.4 | XP_005270108.1 | 2 | 1572 | |||
XM_006717948.3 | XP_006718011.1 | 2 | 743 | |||
XM_006717949.3 | XP_006718012.1 | 2 | 634 | |||
XM_011540091.2 | XP_011538393.1 | 2 | 625 | |||
XM_011540092.2 | XP_011538394.1 | 2 | 634 | |||
XM_011540093.1 | XP_011538395.1 | 2 | 743 | |||
XM_011540094.2 | XP_011538396.1 | 2 | 614 | |||
XM_011540095.2 | XP_011538397.1 | 2 | 614 | |||
XM_017016561.1 | XP_016872050.1 | 2 | 563 | |||
XM_017016562.1 | XP_016872051.1 | 2 | 743 | |||
XM_017016563.1 | XP_016872052.1 | 2 | 556 | |||
XM_017016564.1 | XP_016872053.1 | 2 | 743 | |||
XM_017016565.1 | XP_016872054.1 | 2 | 809 | |||
XM_017016566.1 | XP_016872055.1 | 2 | 634 | |||
XM_017016567.1 | XP_016872056.1 | 2 | 634 | |||
XM_017016568.1 | XP_016872057.1 | 2 | 540 | |||
GenBank mRNA | AF267740.1 | - | 2 | 591 | ||
AK025605.1 | - | 2 | 592 | |||
AK308026.1 | - | 2 | 785 | |||
BC007213.1 | - | 2 | 579 | 2960984 | ||
BC017434.2 | - | 2 | 579 | 2960984 | ||
BX647270.1 | - | 2 | 502 |