Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022157.3 | NP_071440.1 | 1, 2 | 422 | ||
GenBank mRNA | AF272035.1 | - | 1, 2 | 250 | ||
AF323609.1 | - | 1, 2 | 245 | |||
AK315791.1 | - | 1, 2 | 347 | |||
BC016668.1 | - | 1, 2 | 267 | 4342448 | ||
BG717489.1 | - | 1, 2 | 430 |