Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINAGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204414.1 | NP_001191343.1 | 3 | 459 | ||
NM_001204415.1 | NP_001191344.1 | 2 | 67 | |||
NM_022164.2 | NP_071447.1 | 3 | 459 | |||
XM_005271106.2 | XP_005271163.1 | 3 | 459 | |||
XM_005271107.2 | XP_005271164.1 | 2 | 104 | |||
GenBank mRNA | AB050716.1 | - | 3 | 409 | ||
AF205436.1 | - | 3 | 385 | |||
AF236150.1 | - | 3 | 387 | |||
AK074124.1 | - | 2 | 448 | |||
AK075398.1 | - | 3 | 432 | |||
AK292770.1 | - | 3 | 433 | |||
AK296322.1 | - | 3 | 429 | |||
AK298382.1 | - | 3 | 429 | |||
AK301070.1 | - | 3 | 469 | |||
AY358421.1 | - | 3 | 432 | |||
BC064633.1 | - | 3 | 417 | 5750149 | ||
BM923842.1 | - | 2 | 67 | |||
DA656570.1 | - | 3 | 461 |